Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trmt2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080999.2 | NP_001074468.1 | 4 | 1118 | ||
NM_001081000.2 | NP_001074469.1 | 4 | 1165 | |||
NM_001195205.1 | NP_001182134.1 | 4 | 1118 | |||
GenBank mRNA | AK034102.1 | - | 4 | 998 | ||
AK080170.1 | - | 4 | 993 | |||
AK133844.1 | - | 4 | 993 | |||
AK134368.1 | - | 4 | 1119 | |||
AK146391.1 | - | 4 | 1058 | |||
AK150482.1 | - | 4 | 963 | |||
AK151311.1 | - | 4 | 963 | |||
AK151422.1 | - | 4 | 989 | |||
AK151725.1 | - | 4 | 963 | |||
AK151778.1 | - | 4 | 963 | |||
AK152055.1 | - | 4 | 963 | |||
AK152201.1 | - | 4 | 963 | |||
AK152363.1 | - | 4 | 963 | |||
AK152548.1 | - | 4 | 963 | |||
AK152903.1 | - | 4 | 963 | |||
AK153043.1 | - | 4 | 963 | |||
AK153190.1 | - | 4 | 989 | |||
AK169240.1 | - | 4 | 983 | |||
AK182911.1 | - | 1 | 20 | |||
AK185507.1 | - | 1 | 20 | |||
AK206481.1 | - | 1 | 20 | |||
BC027431.1 | - | 3 | 1230 | 3156359 | ||
BC038486.1 | - | 4 | 991 | 3155739 | ||
BC046976.1 | - | 4 | 999 | 6337243 | ||
X56044.1 | - | 3 | 1562 |