Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htr5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008314.2 | NP_032340.2 | 1 | 719 | ||
GenBank mRNA | AK141636.1 | - | 1 | 518 | ||
BC115911.1 | - | 1 | 411 | 40104765 | ||
BC116654.1 | - | 1 | 411 | 40104769 |
Set Membership: |
G-protein coupled receptor |