Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elavl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038698.1 | NP_001033787.1 | 3 | 533 | ||
NM_001163397.1 | NP_001156869.1 | 3 | 369 | |||
NM_001163399.1 | NP_001156871.1 | 3 | 619 | |||
NM_010488.4 | NP_034618.2 | 3 | 619 | |||
GenBank mRNA | AF041341.1 | - | 3 | 381 | ||
AK014133.1 | - | 3 | 387 | |||
AK079088.1 | - | 3 | 524 | |||
BB648788.1 | - | 3 | 620 | |||
BC048159.1 | - | 3 | 592 | 5703005 | ||
BC051987.1 | - | 290 | ||||
BC052451.1 | - | 3 | 496 | 6409658 | ||
CF726962.1 | - | 3 | 369 | |||
CN663544.1 | - | 3 | 533 | |||
D31953.1 | - | 3 | 366 |