Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hus1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF076845.1 | - | 3 | 254 | ||
AK046152.1 | - | 3 | 264 | |||
AK051839.1 | - | 2 | 689 | |||
AK088403.1 | - | 3 | 264 | |||
AK135922.1 | - | 3 | 244 | |||
BB611267.1 | - | 3 | 320 | |||
BC047460.1 | - | 3 | 271 | |||
BC061249.1 | - | 3 | 290 | 30249148 | ||
Y16894.2 | - | 3 | 286 |