Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hus1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF076845.1 | - | 4 | 406 | ||
AK046152.1 | - | 4 | 413 | |||
AK051839.1 | - | 3 | 838 | |||
AK088403.1 | - | 4 | 416 | |||
AK135922.1 | - | 4 | 396 | |||
BB611267.1 | - | 4 | 472 | |||
BC047460.1 | - | 4 | 423 | |||
BC061249.1 | - | 4 | 442 | 30249148 | ||
Y16894.2 | - | 4 | 438 |