Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hyal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010489.2 | NP_034619.2 | 2 | 538 | ||
GenBank mRNA | AF302843.1 | - | 2 | 540 | ||
AF302844.1 | - | 2 | 533 | |||
AF422177.1 | - | 2 | 538 | |||
AF535140.1 | - | 1 | 461 | |||
AJ000059.1 | - | 2 | 564 | |||
AK087358.1 | - | 2 | 560 | |||
AK088806.1 | - | 2 | 576 | |||
AK133886.1 | - | 2 | 584 | |||
BC060047.1 | - | 1 | 766 |