Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Icam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010493.2 | NP_034623.1 | 2, 3 | 397 | ||
GenBank mRNA | AK149748.1 | - | 2, 3 | 373 | ||
AK149781.1 | - | 2, 3 | 372 | |||
AK149945.1 | - | 2, 3 | 372 | |||
AK150003.1 | - | 2, 3 | 373 | |||
AK150049.1 | - | 2, 3 | 372 | |||
AK150057.1 | - | 2, 3 | 373 | |||
AK150141.1 | - | 1, 2 | 110 | |||
AK151227.1 | - | 2, 3 | 373 | |||
AK151681.1 | - | 2, 3 | 373 | |||
AK152155.1 | - | 2, 3 | 373 | |||
AK152527.1 | - | 2, 3 | 373 | |||
AK152530.1 | - | 2, 3 | 373 | |||
AK152556.1 | - | 2, 3 | 373 | |||
AK156417.1 | - | 2, 3 | 373 | |||
AK168275.1 | - | 2, 3 | 373 | |||
AK171502.1 | - | 2, 3 | 373 | |||
AK171520.1 | - | 1, 2 | 110 | |||
AK172321.1 | - | 2, 3 | 373 | |||
CT010246.1 | - | 2, 3 | 322 | |||
M31585.1 | - | 2, 3 | 380 | |||
X16624.1 | - | 2, 3 | 351 | |||
X52264.1 | - | 2, 3 | 344 | |||
X54331.1 | - | 2, 3 | 313 |