Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifi203 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF022371.1 | - | 696 | |||
AK040214.1 | - | 5 | 636 | |||
AK041232.1 | - | 5 | 625 | |||
AK137616.1 | - | 5 | 635 | |||
AK144663.1 | - | 5 | 617 | |||
AK161531.1 | - | 3 | 479 | |||
AK164695.1 | - | 3 | 473 | |||
AK169754.1 | - | 5 | 630 | |||
AK172303.1 | - | 689 | ||||
AK172505.1 | - | 710 | ||||
BC008167.1 | - | 738 | 3257492 | |||
BE632791.1 | - | 3 | 414 | |||
CA574467.1 | - | 3 | 352 | |||
L14559.1 | - | 1 | 128 |