Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifi47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271676.1 | NP_001258605.1 | 3 | 1322 | ||
NM_001271677.1 | NP_001258606.1 | 3 | 1171 | |||
NM_008330.2 | NP_032356.1 | 2 | 1112 | |||
GenBank mRNA | AK156785.1 | - | 2 | 1091 | ||
BC001986.1 | - | 2 | 1079 | 3589815 | ||
BC009131.1 | - | 2 | 1069 | 2651113 | ||
CT010353.1 | - | 1 | 965 | |||
M63630.1 | - | 2 | 1074 |