Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifi47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271676.1 | NP_001258605.1 | 3 | 441 | ||
NM_001271677.1 | NP_001258606.1 | 3 | 290 | |||
NM_008330.2 | NP_032356.1 | 2 | 231 | |||
GenBank mRNA | AK156785.1 | - | 2 | 210 | ||
BC001986.1 | - | 2 | 198 | 3589815 | ||
BC009131.1 | - | 2 | 188 | 2651113 | ||
BU696734.1 | - | 3 | 290 | |||
BY159742.1 | - | 2 | 232 | |||
CT010353.1 | - | 1 | 84 | |||
M63630.1 | - | 2 | 193 |