Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004502.3 | NP_004493.3 | 2 | 570 | ||
GenBank mRNA | AK290653.1 | - | 2 | 536 | ||
BC015345.1 | - | 2 | 517 | 4413080 | ||
BG118354.1 | - | 568 | ||||
BM923808.1 | - | 2 | 571 | |||
M30598.1 | - | 2 | 113 |