Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifit2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008332.3 | NP_032358.1 | 3 | 1310 | ||
GenBank mRNA | AK038027.1 | - | 3 | 1063 | ||
AK042366.1 | - | 3 | 1092 | |||
AK150206.1 | - | 2 | 912 | |||
AK150253.1 | - | 2 | 912 | |||
BC050835.1 | - | 3 | 1058 | 6513417 | ||
CT010395.1 | - | 1 | 834 | |||
U43085.1 | - | 2 | 885 |