Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifna2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010503.2 | NP_034633.2 | 1 | 240 | ||
GenBank mRNA | BC100688.1 | - | 1 | 238 | ||
BC100689.1 | - | 1 | 239 | |||
BC100698.1 | - | 1 | 239 | |||
BC100699.1 | - | 1 | 239 | |||
BC114423.1 | - | 1 | 233 | |||
K01411.1 | - | 1 | 78 |
Gene Symbol: Ifna11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008333.2 | NP_032359.2 | 1 | 280 | ||
GenBank mRNA | BC116870.1 | - | 1 | 280 | 40126189 | |
BC116872.1 | - | 1 | 280 | 40126191 |