Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004502.3 | NP_004493.3 | 1, 2 | 506 | ||
GenBank mRNA | AK223249.1 | - | 1, 2 | 505 | ||
AK290653.1 | - | 1, 2 | 472 | |||
BC015345.1 | - | 1, 2 | 453 | 4413080 | ||
BG118354.1 | - | 507 | ||||
BM923808.1 | - | 1, 2 | 507 | |||
M16937.1 | - | 1, 2 | 502 | |||
M30598.1 | - | 1, 2 | 49 | |||
S49765.1 | - | 1, 2 | 112 |