Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifnar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010508.2 | NP_034638.2 | 3 | 478 | ||
GenBank mRNA | AK132431.1 | - | 3 | 410 | ||
AK136950.1 | - | 3 | 455 | |||
AK141630.1 | - | 3 | 414 | |||
AK148246.1 | - | 3 | 420 | |||
AK157555.1 | - | 3 | 445 | |||
BC043935.1 | - | 3 | 429 | 4507009 | ||
BC052217.1 | - | 3 | 438 | 30008497 | ||
BC052429.1 | - | 3 | 440 | 6836300 | ||
M89641.1 | - | 3 | 443 |