Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igf2r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010515.2 | NP_034645.2 | 23 | 3274 | ||
GenBank mRNA | BC150817.1 | - | 23 | 3185 | 9087728 | |
BC171973.1 | - | 23 | 3185 | 9054259 | ||
CF167933.1 | - | 1 | 36 | |||
U04710.1 | - | 23 | 3230 |