Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il10ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008348.2 | NP_032374.1 | 3 | 350 | ||
GenBank mRNA | AK137663.1 | - | 3 | 351 | ||
AK149680.1 | - | 3 | 345 | |||
AK149899.1 | - | 2 | 233 | |||
AK150022.1 | - | 3 | 347 | |||
AK150524.1 | - | 3 | 354 | |||
AK153120.1 | - | 3 | 354 | |||
AK159566.1 | - | 3 | 347 | |||
L12120.1 | - | 3 | 355 |