Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il10rb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008349.5 | NP_032375.2 | 4 | 573 | ||
GenBank mRNA | AF440787.1 | - | 4 | 500 | ||
AK040938.1 | - | 4 | 489 | |||
AK134042.1 | - | 4 | 566 | |||
AK160991.1 | - | 4 | 1528 | |||
BC145791.1 | - | 4 | 573 | 40131094 | ||
U53696.1 | - | 4 | 486 |