Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il15ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271497.1 | NP_001258426.1 | 7 | 809 | ||
NM_001271498.1 | NP_001258427.1 | 7 | 903 | |||
NM_001271499.1 | NP_001258428.1 | 7 | 773 | |||
NM_001271500.1 | NP_001258429.1 | 6 | 674 | |||
NM_001271501.1 | NP_001258430.1 | 5 | 581 | |||
NM_008358.2 | NP_032384.1 | 7 | 872 | |||
NM_133836.2 | NP_598597.1 | 7 | 925 | |||
GenBank mRNA | AK040940.1 | - | 7 | 755 | ||
AK080000.1 | - | 7 | 755 | |||
AY219715.1 | - | 7 | 674 | |||
AY219716.1 | - | 6 | 576 | |||
AY219717.1 | - | 5 | 483 | |||
AY221616.1 | - | 7 | 550 | |||
AY221617.1 | - | 6 | 468 | |||
AY221618.1 | - | 5 | 375 | |||
AY221619.1 | - | 7 | 642 | |||
BC022705.1 | - | 7 | 682 | |||
BC069881.1 | - | 6 | 633 | |||
BC095982.1 | - | 4 | 600 | |||
BC132231.1 | - | 6 | 472 | |||
BC132233.1 | - | 6 | 472 | |||
BC145348.1 | - | 7 | 543 |