Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004503.3 | NP_004494.1 | 1 | 327 | ||
NM_153693.4 | NP_710160.1 | 2 | 724 | |||
GenBank mRNA | BC074844.2 | - | 1 | 241 | 30915460 | |
BC074845.2 | - | 1 | 241 | 30915223 | ||
CR456954.1 | - | 1 | 215 | |||
M16938.1 | - | 2 | 451 | |||
S82986.1 | - | 1 | 347 |