Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008360.1 | NP_032386.1 | 4 | 367 | ||
GenBank mRNA | AK078100.1 | - | 5 | 518 | ||
AY157834.1 | - | 4 | 360 | |||
AY157835.1 | - | 4 | 360 | |||
AY362457.1 | - | 3 | 212 | |||
AY916131.1 | - | 3 | 203 | |||
BC024384.1 | - | 4 | 361 | 4988988 | ||
D49949.1 | - | 4 | 367 | |||
DQ316604.1 | - | 3 | 203 | |||
U66244.1 | - | 3 | 203 |