Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008361.3 | NP_032387.1 | 5 | 454 | ||
GenBank mRNA | AK156396.1 | - | 4 | 1006 | ||
AK168047.1 | - | 5 | 474 | |||
AK224980.1 | - | 4 | 1006 | |||
AK225002.1 | - | 5 | 473 | |||
AK225003.1 | - | 1 | 29 | |||
BC011437.1 | - | 5 | 454 | 3989461 | ||
M15131.1 | - | 5 | 463 |