Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il1r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123382.1 | NP_001116854.1 | 8 | 1222 | ||
NM_008362.2 | NP_032388.1 | 8 | 1153 | |||
GenBank mRNA | AK048550.1 | - | 8 | 1140 | ||
AK054137.1 | - | 8 | 1223 | |||
BC109135.2 | - | 8 | 1059 | 40039321 | ||
M20658.1 | - | 8 | 1153 |