Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008366.3 | NP_032392.1 | 3 | 320 | ||
GenBank mRNA | AF065914.1 | - | 3 | 318 | ||
AF065915.1 | - | 3 | 318 | |||
AF065916.1 | - | 3 | 318 | |||
AY147902.1 | - | 3 | 272 | |||
BC116845.1 | - | 3 | 318 | 40126164 | ||
BC116873.1 | - | 3 | 318 | 40126192 | ||
DQ836354.1 | - | 3 | 212 | |||
K02292.1 | - | 3 | 320 | |||
U41494.1 | - | 3 | 215 | |||
U41504.1 | - | 3 | 230 | |||
U41505.1 | - | 3 | 242 | |||
U41506.1 | - | 3 | 215 | |||
X01772.1 | - | 3 | 319 | |||
X73040.1 | - | 3 | 319 |