Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il3ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008369.1 | NP_032395.1 | 6 | 710 | ||
GenBank mRNA | BC049889.1 | - | 5 | 1110 | ||
FJ550346.1 | - | 3 | 292 | |||
X64534.1 | - | 6 | 710 |