Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il3ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008369.1 | NP_032395.1 | 3 | 236 | ||
GenBank mRNA | BC049889.1 | - | 2 | 636 | ||
BC077736.1 | - | 2 | 177 | |||
X64534.1 | - | 3 | 236 |