Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008371.4 | NP_032397.1 | 4 | 586 | ||
GenBank mRNA | AK040399.1 | - | 4 | 370 | ||
AK041307.1 | - | 4 | 587 | |||
AK041403.1 | - | 288 | ||||
AK042009.1 | - | 6 | 1371 | |||
AK052452.1 | - | 5 | 1274 | |||
AK139858.1 | - | 5 | 947 | |||
AK145184.1 | - | 4 | 877 | |||
BC110553.2 | - | 4 | 495 | 40047584 | ||
X07962.1 | - | 4 | 886 |