Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep250 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129999.1 | NP_001123471.1 | 6, 7 | 799 | ||
NM_001130000.1 | NP_001123472.1 | 6, 7 | 794 | |||
NM_008383.3 | NP_032409.3 | 6, 7 | 799 | |||
NM_177217.3 | NP_796191.2 | 6, 7 | 794 | |||
GenBank mRNA | AK033281.1 | - | 6, 7 | 763 | ||
AK045557.1 | - | 6, 7 | 786 | |||
AK145562.1 | - | 6, 7 | 795 | |||
AK161443.1 | - | 6, 7 | 900 | |||
BC068153.1 | - | 6, 7 | 782 | |||
BC145268.1 | - | 5, 6 | 621 | |||
BC150746.1 | - | 5, 6 | 621 | 9087657 | ||
DQ148475.1 | - | 5, 6 | 594 |