Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inpp5d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110192.2 | NP_001103662.1 | 9 | 1156 | ||
NM_001110193.2 | NP_001103663.1 | 9 | 1159 | |||
NM_010566.3 | NP_034696.2 | 9 | 1159 | |||
GenBank mRNA | AF125996.1 | - | 9 | 1072 | ||
AF184912.1 | - | 4 | 352 | |||
AF184913.1 | - | 4 | 352 | |||
AF228679.1 | - | 9 | 1070 | |||
AK143560.1 | - | 9 | 1096 | |||
BC108328.1 | - | 9 | 1032 | 30839591 | ||
U39203.1 | - | 9 | 1108 | |||
U51742.1 | - | 9 | 1070 | |||
U52044.1 | - | 9 | 1092 |