Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif3e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008388.2 | NP_032414.1 | 4 | 384 | ||
GenBank mRNA | AK002576.1 | - | 4 | 361 | ||
AK011436.1 | - | 4 | 354 | |||
AK088104.1 | - | 4 | 363 | |||
AK088269.1 | - | 4 | 359 | |||
AK145059.1 | - | 4 | 367 | |||
AK146936.1 | - | 4 | 356 | |||
AK161650.1 | - | 4 | 354 | |||
AK165929.1 | - | 4 | 362 | |||
BC010252.1 | - | 4 | 367 | |||
BC029177.1 | - | 4 | 345 | 3988386 | ||
U54563.1 | - | 4 | 350 |