Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010574.2 | NP_034704.1 | 3 | 1279 | ||
GenBank mRNA | AF165986.1 | - | 3 | 1183 | ||
AF295369.1 | - | 3 | 1195 | |||
AK018266.1 | - | 2 | 840 | |||
AK148520.1 | - | 3 | 1438 | |||
BC029750.1 | - | 3 | 1279 | 3592499 | ||
BC085291.1 | - | 4 | 1291 | 5347048 |