Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253822.1 | NP_001240751.1 | 3 | 1862 | ||
NM_008393.3 | NP_032419.2 | 3 | 1848 | |||
GenBank mRNA | AK143023.1 | - | 3 | 1764 | ||
BC067195.1 | - | 3 | 1847 | |||
BC085500.1 | - | 3 | 1568 | 30539191 | ||
Y15001.1 | - | 3 | 1434 |