Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159417.1 | NP_001152889.1 | 7 | 935 | ||
NM_001159418.1 | NP_001152890.1 | 6 | 733 | |||
NM_008394.3 | NP_032420.1 | 6 | 849 | |||
GenBank mRNA | AK043697.1 | - | 3 | 408 | ||
AK145911.1 | - | 5 | 601 | |||
AK149742.1 | - | 5 | 600 | |||
BC005435.1 | - | 7 | 710 | 3601255 | ||
BC012968.1 | - | 6 | 744 | 3257714 | ||
CK788642.1 | - | 6 | 676 | |||
U51992.1 | - | 6 | 653 |