Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243712.1 | NP_001230641.1 | 20 | 2266 | ||
NM_008395.3 | NP_032421.2 | 19 | 2064 | |||
GenBank mRNA | AF037454.1 | - | 19 | 1980 | ||
AK048303.1 | - | 19 | 1985 | |||
AK220164.1 | - | 20 | 2188 | |||
BC062934.1 | - | 19 | 2013 | 30535756 | ||
BC064678.1 | - | 19 | 1981 | 6409535 |