Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itga5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010577.3 | NP_034707.3 | 28 | 3110 | ||
GenBank mRNA | AK142241.1 | - | 28 | 2953 | ||
AK144921.1 | - | 28 | 2897 | |||
AK148138.1 | - | 28 | 2951 | |||
AK199283.1 | - | 1 | 35 | |||
BC050943.1 | - | 28 | 3126 | 6821577 | ||
BC053431.1 | - | 29 | 3159 | |||
BC059829.1 | - | 28 | 3126 | 6821577 | ||
X15203.1 | - | 10 | 956 | |||
X79003.1 | - | 28 | 3067 |