Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itga7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008398.2 | NP_032424.2 | 4 | 611 | ||
GenBank mRNA | AK157612.1 | - | 4 | 606 | ||
BQ933931.1 | - | 4 | 377 | |||
L23423.1 | - | 4 | 609 |