Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253872.1 | NP_001240801.1 | 5 | 561 | ||
NM_001253873.1 | NP_001240802.1 | 5 | 561 | |||
NM_001253874.1 | NP_001240803.1 | 5 | 561 | |||
NM_008400.3 | NP_032426.2 | 5 | 561 | |||
GenBank mRNA | AF065901.1 | - | 5 | 363 | ||
AF065902.1 | - | 5 | 363 | |||
AK152913.1 | - | 5 | 361 | |||
AK154115.1 | - | 5 | 496 | |||
AK155566.1 | - | 5 | 445 | |||
AK156251.1 | - | 5 | 531 | |||
AK169628.1 | - | 5 | 477 | |||
AK171389.1 | - | 5 | 445 | |||
AK172394.1 | - | 5 | 507 | |||
BC145094.1 | - | 5 | 375 | 9053642 | ||
BC145095.1 | - | 5 | 375 | 9053643 | ||
BC145096.1 | - | 5 | 375 | 9053644 | ||
BC145802.1 | - | 5 | 375 | 40131105 | ||
JN986841.1 | - | 5 | 356 | |||
M60778.1 | - | 5 | 411 |