Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253872.1 | NP_001240801.1 | 6 | 709 | ||
NM_001253873.1 | NP_001240802.1 | 6 | 709 | |||
NM_001253874.1 | NP_001240803.1 | 6 | 709 | |||
NM_008400.3 | NP_032426.2 | 6 | 709 | |||
GenBank mRNA | AF065901.1 | - | 6 | 511 | ||
AF065902.1 | - | 6 | 511 | |||
AK152913.1 | - | 6 | 509 | |||
AK154115.1 | - | 6 | 644 | |||
AK155566.1 | - | 6 | 593 | |||
AK156251.1 | - | 6 | 679 | |||
AK169628.1 | - | 6 | 625 | |||
AK171389.1 | - | 6 | 593 | |||
AK172394.1 | - | 6 | 655 | |||
BC145094.1 | - | 6 | 523 | 9053642 | ||
BC145095.1 | - | 6 | 523 | 9053643 | ||
BC145096.1 | - | 6 | 523 | 9053644 | ||
BC145802.1 | - | 6 | 523 | 40131105 | ||
JN986841.1 | - | 6 | 504 | |||
M60778.1 | - | 6 | 559 |