Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159564.1 | NP_001153036.1 | 9 | 1196 | ||
NM_021359.3 | NP_067334.1 | 6 | 1090 | |||
GenBank mRNA | AF115376.1 | - | 9 | 1042 | ||
AK008867.1 | - | 6 | 1089 | |||
AK019511.1 | - | 6 | 953 | |||
AK036439.1 | - | 9 | 1197 | |||
AK085900.1 | - | 9 | 1237 | |||
BC049185.1 | - | 6 | 1059 | 6475423 |