Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itih2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010582.3 | NP_034712.2 | 4, 5 | 485 | ||
GenBank mRNA | AK133782.1 | - | 4, 5 | 384 | ||
AK149533.1 | - | 5, 6 | 530 | |||
BC034341.1 | - | 4, 5 | 459 | 4211576 | ||
X70392.1 | - | 4, 5 | 471 |