Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281965.1 | NP_001268894.1 | 16 | 1791 | ||
NM_001281966.1 | NP_001268895.1 | 16 | 1782 | |||
NM_001281968.1 | NP_001268897.1 | 16 | 1770 | |||
NM_010583.3 | NP_034713.2 | 16 | 1773 | |||
GenBank mRNA | AK037554.1 | - | 853 | |||
AK083380.1 | - | 16 | 1745 | |||
AK153613.1 | - | 16 | 1729 | |||
BC128374.1 | - | 16 | 1699 | 40111687 | ||
BC128375.1 | - | 16 | 1696 | 40111688 | ||
D14042.1 | - | 16 | 1713 | |||
L00619.1 | - | 16 | 1778 | |||
L05631.1 | - | 16 | 1739 | |||
L10628.1 | - | 16 | 1741 |