Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010584.3 | NP_034714.1 | 7 | 798 | ||
GenBank mRNA | AB016496.1 | - | 7 | 797 | ||
AK007447.1 | - | 7 | 798 | |||
AY157363.1 | - | 7 | 787 | |||
AY157364.1 | - | 7 | 782 | |||
AY619693.1 | - | 7 | 781 | |||
BC145377.1 | - | 7 | 744 | 9053935 | ||
BC150760.1 | - | 7 | 744 | 9087671 | ||
BC150795.1 | - | 7 | 740 | 9087706 |