Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itpr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010585.5 | NP_034715.3 | 38, 39 | 5306 | ||
GenBank mRNA | AK087694.1 | - | 1, 2 | 1857 | ||
CB521690.1 | - | 1, 2 | 159 | |||
CF168791.1 | - | 2, 3 | 297 | |||
CF744794.1 | - | 4, 5 | 571 | |||
X15373.1 | - | 38, 39 | 5284 |