Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146145.2 | NP_666257.2 | 9 | 1613 | ||
GenBank mRNA | AK133866.1 | - | 9 | 1556 | ||
AK141210.1 | - | 9 | 1555 | |||
AK143650.1 | - | 9 | 1559 | |||
AK148521.1 | - | 9 | 1577 | |||
AK152128.1 | - | 9 | 1576 | |||
BC024100.1 | - | 1 | 23 | |||
S63728.1 | - | 9 | 1563 |