Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atcay | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178662.3 | NP_848777.1 | 8, 9 | 1213 | ||
GenBank mRNA | AK028298.1 | - | 8, 9 | 1220 | ||
AK038483.1 | - | 8, 9 | 1223 | |||
AK038990.1 | - | 8, 9 | 1214 | |||
AK039411.1 | - | 8, 9 | 1221 | |||
AK086723.1 | - | 8, 9 | 1256 | |||
AK162959.1 | - | 8, 9 | 1221 | |||
AY349150.1 | - | 8, 9 | 1178 | |||
BC048903.1 | - | 8, 9 | 1197 | 6491141 |