Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jarid2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205043.1 | NP_001191972.1 | 10 | 3155 | ||
NM_001205044.1 | NP_001191973.1 | 11 | 2828 | |||
NM_021878.3 | NP_068678.1 | 11 | 2874 | |||
GenBank mRNA | AK045214.1 | - | 10 | 2960 | ||
AK147226.1 | - | 10 | 3155 | |||
AK164134.1 | - | 9 | 2479 | |||
BC003374.1 | - | 4 | 724 | |||
BC026056.1 | - | 10 | 3062 | |||
BC050129.1 | - | 10 | 3076 | |||
BC051010.1 | - | 10 | 3058 | |||
BC052444.1 | - | 11 | 2745 | 6406875 | ||
BC060695.1 | - | 11 | 2745 | 6406875 | ||
BC115758.1 | - | 5 | 1759 | |||
D31967.1 | - | 11 | 2764 |