Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010595.3 | NP_034725.3 | 2 | 3496 | ||
GenBank mRNA | AK039559.1 | - | 2 | 2491 | ||
BC112970.2 | - | 2 | 2101 | 40048563 | ||
Y00305.1 | - | 1 | 2875 |