Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcna6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013568.6 | NP_038596.1 | 1 | 2290 | ||
GenBank mRNA | BC048782.1 | - | 1 | 1772 | 6466414 | |
BC054804.1 | - | 1 | 1810 | 6838056 | ||
M96688.1 | - | 1 | 2455 |