Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008420.4 | NP_032446.2 | 2 | 811 | ||
GenBank mRNA | BC031776.1 | - | 2 | 767 | 4507847 | |
BC051422.1 | - | 2 | 768 | 6493266 | ||
BC061501.1 | - | 2 | 792 | 4506061 | ||
DQ646376.1 | - | 1065 | ||||
DQ646377.1 | - | 2 | 811 | |||
M64228.1 | - | 2 | 616 |