Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013569.2 | NP_038597.2 | 5 | 1402 | ||
GenBank mRNA | AF012868.1 | - | 5 | 1408 | ||
AY374424.1 | - | 5 | 1087 | |||
BC051016.1 | - | 5 | 1233 | |||
BQ745654.1 | - | 2 | 296 |